2-Oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Oxazolidone, Oxazolidin-2-one

M-Clarity™ Qualitätsstufe = MQ100

O9409-100G O9409-25G O9409-5G
SIALO9409-100GEA 111 EUR
SIALO9409-100G SIALO9409-25G SIALO9409-5G
2-Oxazolidinon, Sigma-Aldrich®
2-Oxazolidinon
Formel: C₃H₅NO₂
Molecular Weight: 87,08 g/mol
MDL: MFCD00005268
CAS-Nummer: 497-25-6
EINECS: 207-840-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR