3-(Trimethylsilyl)-2-oxazolidinon

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Trimethylsilyl)oxazolidin-2-one

SILS19925.5 SILS19925.25
SILNSILS19925.5EA 0 EUR
SILNSILS19925.5 SILNSILS19925.25
3-(Trimethylsilyl)-2-oxazolidinon
3-(Trimethylsilyl)-2-oxazolidinon
Formel: C₆H₁₃NO₂Si
Molecular Weight: 159.26 g/mol
MDL: MFCD00010197
CAS-Nummer: 43112-38-5
EINECS: 256-098-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR