6-Brom-2-tetralon 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
6-Bromo-3,4-dihydronaphthalen-2(1H)-one
330770010
330770050
ACRO330770010EA
50.1
EUR
ACRO330770010
ACRO330770050
6-Brom-2-tetralon 98%
6-Brom-2-tetralon
Formel:
C₁₀H₉BrO Molecular Weight: 225.08 g/mol Schmelzpunkt: 70…76 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00239388 CAS-Nummer: 4133-35-1 |
Spezifikation Testergebnisse
Appearance | Light yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 70 to 76 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...