7-Brom-1-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

708453-10G 708453-1G
SIAL708453-10GEA 158 EUR
SIAL708453-10G SIAL708453-1G
7-Brom-1-tetralon, Sigma-Aldrich®
7-Brom-1-tetralon
Formel: C₁₀H₉BrO
Molecular Weight: 225,08 g/mol
MDL: MFCD02179287
CAS-Nummer: 32281-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR