6-Methoxy-1-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone, 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one

M-Clarity™ Qualitätsstufe = MQ100

M25589-100G M25589-25G
SIALM25589-100GEA 85.9 EUR
SIALM25589-100G SIALM25589-25G
6-Methoxy-1-tetralon, Sigma-Aldrich®
6-Methoxy-1-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD00001695
CAS-Nummer: 1078-19-9
EINECS: 214-078-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR