6-Methoxy-2-tetralon

Lieferant: CHEMPUR

Synonyme: 6-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-6-methoxynaphthalen-2(1H)-one

10-0004.5 10-0004.10
CHMP10-0004.5EA 0 EUR
CHMP10-0004.5 CHMP10-0004.10
6-Methoxy-2-tetralon
6-Methoxy-2-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD00001729
CAS-Nummer: 2472-22-2
EINECS: 219-592-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR