8-Methoxy-2-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 8-Methoxy-3,4-dihydronaphthalen-2(1H)-one

M-Clarity™ Qualitätsstufe = MQ100

535451-1G 535451-5G
SIAL535451-1GEA 177 EUR
SIAL535451-1G SIAL535451-5G
8-Methoxy-2-tetralon, Sigma-Aldrich®
8-Methoxy-2-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD06411220
CAS-Nummer: 5309-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR