6-Fluor-2-tetralon

Lieferant: CHEMPUR

Synonyme: 6-Fluoro-3,4-dihydronaphthalen-2(1H)-one

10-0007.1 10-0007.5
CHMP10-0007.1EA 0 EUR
CHMP10-0007.1 CHMP10-0007.5
6-Fluor-2-tetralon
6-Fluor-2-tetralon
Formel: C₁₀H₉FO
Molecular Weight: 164,18 g/mol
MDL: MFCD02684435
CAS-Nummer: 29419-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR