7-Fluor-1-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Fluoro-3,4-dihydronaphthalen-1(2H)-one

722251-1G 722251-5G
SIAL722251-1GEA 43.2 EUR
SIAL722251-1G SIAL722251-5G
7-Fluor-1-tetralon, Sigma-Aldrich®
7-Fluor-1-tetralon
Formel: C₁₀H₉FO
Molecular Weight: 164,18 g/mol
MDL: MFCD07375433
CAS-Nummer: 2840-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR