5,6,7,8-Tetrahydro-1-naphthalinol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
L02416.09
L02416.18
L02416.09EA
101
EUR
L02416.09
L02416.18
5,6,7,8-Tetrahydro-1-naphthalinol ≥99%
5,6,7,8-Tetrahydro-1-naphthol
Purity: 99%
CAS No.: 529-35-1
Molecular Formula: C10H12O
Formula Weight: 148.21
Melting Point: 69-71°
Boiling Point: 264-265°/705mm
MDL No.: MFCD00001734
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 264…265 °C (705 mmHg) Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001734 CAS-Nummer: 529-35-1 EINECS: 208-461-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...