5,6,7,8-Tetrahydro-2-naphthalinol

Lieferant: COMBI-BLOCKS

OS-7715-5G OS-7715-25G OS-7715-100G
COBBOS-7715-5GEA 48.4 EUR
COBBOS-7715-5G COBBOS-7715-25G COBBOS-7715-100G
5,6,7,8-Tetrahydro-2-naphthalinol
5,6,7,8-Tetrahydro-2-naphthol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
MDL: MFCD00001738
CAS-Nummer: 1125-78-6
EINECS: 214-410-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR