(2E,4E)-2,4-Hexadienal, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W342904-100G W342904-1KG W342904-4KG
SIALW342904-100GEA 120 EUR
SIALW342904-100G SIALW342904-1KG SIALW342904-4KG
(2E,4E)-2,4-Hexadienal, Sigma-Aldrich®
trans,trans-2,4-Hexadienal
Formel: C₆H₈O
MDL: MFCD00007004
CAS-Nummer: 142-83-6
EINECS: 205-564-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR