(2E,4E)-2,4-Undecadienal, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W342203-100G-K W342203-1KG-K W342203-25G-K
SIALW342203-100G-KEA 360 EUR
SIALW342203-100G-K SIALW342203-1KG-K SIALW342203-25G-K
(2E,4E)-2,4-Undecadienal, Sigma-Aldrich®
trans,trans-2,4-Undecadienal
Formel: C₁₁H₁₈O
MDL: MFCD00014677
CAS-Nummer: 30361-29-6
EINECS: 250-148-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR