(2E,6E)-2,6-Nonadienal, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W376604-100G-K W376604-25G-K
SIALW376604-100G-KEA 1130 EUR
SIALW376604-100G-K SIALW376604-25G-K
(2E,6E)-2,6-Nonadienal, Sigma-Aldrich®
trans,trans-2,6-Nonadienal
Formel: C₉H₁₄O
Molecular Weight: 138,21 g/mol
MDL: MFCD00209523
CAS-Nummer: 17587-33-6
EINECS: 241-557-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR