Ketoprofen, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid
M-Clarity™ Qualitätsstufe = MQ200
K2012-5G
K2012-25G
SIALK2012-5GEA
140
EUR
SIALK2012-5G
SIALK2012-25G
Ketoprofen, Sigma-Aldrich®
Ketoprofen
Formel:
C₁₆H₁₄O₃ Molecular Weight: 254,29 g/mol |
MDL:
MFCD00055790 CAS-Nummer: 22071-15-4 EINECS: 244-759-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...