N-(Benzyloxycarbonyloxy)succinimid 98%
Lieferant: Thermo Scientific
Synonyme:
CBZ-OSu (Z-OSu)
211170250
211171000
211175000
ACRO211170250EA
51.3
EUR
ACRO211170250
ACRO211171000
ACRO211175000
N-(Benzyloxycarbonyloxy)succinimid 98%
N-(Benzyloxycarbonyloxy)succinimid
Formel:
C₁₂H₁₁NO₅ Molecular Weight: 249,22 g/mol Schmelzpunkt: 77…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005513 CAS-Nummer: 13139-17-8 EINECS: 236-075-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 82 °C |
Loss on drying | ≤0.25 % (60 °C, 2 h, 1 mmHg) |
Solubility | (10 % in acetone) Clear colorless to almost colorless |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...