N-(Benzyloxycarbonyloxy)succinimid ≥98%

Lieferant: Thermo Scientific

Synonyme: CBZ-OSu (Z-OSu)

A12153.14 A12153.22
A12153.14EA 54.1 EUR
A12153.14 A12153.22
N-(Benzyloxycarbonyloxy)succinimid ≥98%
N-(Benzyloxycarbonyloxy)succinimid

Purity: 99% CAS No.: 13139-17-8 Molecular Formula: C12H11NO5 Formula Weight: 249.23 Storage Temperature: 2°C to 8°C Melting Point: 77-82° Beilstein Reference: 1387927 MDL No.: MFCD00005513

Formel: C₁₂H₁₁NO₅
Molecular Weight: 249,22 g/mol
Schmelzpunkt: 77…82 °C
MDL: MFCD00005513
CAS-Nummer: 13139-17-8
EINECS: 236-075-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR