N-(Benzyloxycarbonyloxy)succinimid

Lieferant: BACHEM BIOCHEMICA

Synonyme: CBZ-OSu (Z-OSu)

4031237.0100 4031237.0500
BACE4031237.0100EA 0 EUR
BACE4031237.0100 BACE4031237.0500
N-(Benzyloxycarbonyloxy)succinimid
N-(Benzyloxycarbonyloxy)succinimid
Formel: C₁₂H₁₁NO₅
Molecular Weight: 249.22 g/mol
MDL: MFCD00005513
CAS-Nummer: 13139-17-8
EINECS: 236-075-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR