Phenylarsinoxid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: PAO

180350050 180350010 180350250
ACRO180350050EA 77.9 EUR
ACRO180350050 ACRO180350010 ACRO180350250
Phenylarsinoxid 97%
Phenylarsine oxide
Formel: C₆H₅AsO
Molecular Weight: 168.02 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001990
CAS-Nummer: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1 II

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥96.0 %
HPLC ≥98.5 %
Optical rotation +24° to +28° (1 % in DMF/methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR