Phenylarsinoxid 97

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: PAO

A14674.03 A14674.06 A14674.14
A14674.03EA 31.6 EUR
A14674.03 A14674.06 A14674.14
Phenylarsinoxid 97
Phenylarsine oxide

Purity: 97% CAS No.: 637-03-6 Molecular Formula: C6H5AsO Formula Weight: 168.03 Melting Point: 140-160° Beilstein Reference: 2935227 Merck Reference: 14,6949 UN No.: 3465 MDL No.: MFCD00001990

Formel: C₆H₅AsO
Molecular Weight: 168.02 g/mol
Schmelzpunkt: 140…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001990
CAS-Nummer: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR