Phenylarsinoxid ≥97%, Pulver, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: PAO

M-Clarity™ Qualitätsstufe = MQ100

P3075-5G P3075-1G
SIALP3075-5GEA 290 EUR
SIALP3075-5G SIALP3075-1G
Phenylarsinoxid ≥97%, Pulver, Sigma-Aldrich®
Phenylarsine oxide
Formel: C₆H₅AsO
Molecular Weight: 168.02 g/mol
MDL: MFCD00001990
CAS-Nummer: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR