3-Methyl-2(5H)-furanon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Hydroxy-2-methyl-2-butenoic acid gamma-Lactone
M-Clarity™ Qualitätsstufe = MQ100
393509-1G
393509-5G
SIAL393509-1GEA
197
EUR
SIAL393509-1G
SIAL393509-5G
3-Methyl-2(5H)-furanon, Sigma-Aldrich®
3-Methyl-2(5H)-furanon
Formel:
C₅H₆O₂ Molecular Weight: 98,1 g/mol |
MDL:
MFCD00191545 CAS-Nummer: 22122-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...