3-Chlor-6-hydrazinopyridazin
Lieferant: Thermo Scientific
Synonyme:
3-Chloro-6-hydrazinylpyridazine
BTB05871.1
BTB05871.10
MAYBBTB05871.1EA
0
EUR
MAYBBTB05871.1
MAYBBTB05871.10
3-Chlor-6-hydrazinopyridazin
3-Chlor-6-hydrazinopyridazin
Formel:
C₄H₅ClN₄ Molecular Weight: 144,56 g/mol |
MDL:
MFCD00051740 CAS-Nummer: 17284-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...