2-Pyrimidincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Cyanopyrimidine

M-Clarity™ Qualitätsstufe = MQ100

646830-25G 646830-5G
SIAL646830-25GEA 77.4 EUR
SIAL646830-25G SIAL646830-5G
2-Pyrimidincarbonitril, Sigma-Aldrich®
2-Pyrimidincarbonitril
Formel: C₅H₃N₃
Molecular Weight: 105.1 g/mol
MDL: MFCD00160513
CAS-Nummer: 14080-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR