2-Methoxypyrimidin

Lieferant: COMBI-BLOCKS

PY-7838-1G PY-7838-5G
COBBPY-7838-1GEA 303 EUR
COBBPY-7838-1G COBBPY-7838-5G
2-Methoxypyrimidin
2-Methoxypyrimidine
Formel: C₅H₆N₂O
MDL: MFCD00234104
CAS-Nummer: 931-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR