1,4-Dithian-2,5-diol
Lieferant: FLUOROCHEM
Synonyme:
2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol
022124-100G
022124-1KG
FLUO022124-100GEA
84.9
EUR
FLUO022124-100G
FLUO022124-1KG
1,4-Dithian-2,5-diol
1,4-Dithian-2,5-diol
Formel:
C₄H₈O₂S₂ Molecular Weight: 152,24 g/mol |
MDL:
MFCD00006659 CAS-Nummer: 40018-26-6 EINECS: 254-751-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...