1,4-Dithian-2,5-diol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol

M-Clarity™ Qualitätsstufe = MQ100

183954-250G 183954-50G
SIAL183954-250GEA 103 EUR
SIAL183954-250G SIAL183954-50G
1,4-Dithian-2,5-diol, Sigma-Aldrich®
1,4-Dithian-2,5-diol
Formel: C₄H₈O₂S₂
Molecular Weight: 152,24 g/mol
MDL: MFCD00006659
CAS-Nummer: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR