N-Acetyl-L-glutaminsäure ≥99%
Lieferant: Thermo Scientific
B23621.14
B23621.22
B23621.14EA
28.6
EUR
B23621.14
B23621.22
N-Acetyl-L-glutaminsäure ≥99%
N-Acetyl-L-glutaminsäure
Purity: 99%
CAS No.: 1188-37-0
Molecular Formula: C7H11NO5
Formula Weight: 189.17
Storage Temperature: 2°C to 8°C
Melting Point: 194-199°
Beilstein Reference: 1727473
Optical Rotation: -16.5° (c=1 in water)
MDL No.: MFCD00002802
Formel:
C₇H₁₁NO₅ Molecular Weight: 189,17 g/mol Schmelzpunkt: 194…199 °C |
MDL:
MFCD00002802 CAS-Nummer: 1188-37-0 EINECS: 214-708-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...