Hexafluoraceton Sesquihydrat

Lieferant: FLUOROCHEM

Synonyme: 1,1,1,3,3,3-Hexafluoroacetone sesquihydrate

003250-25G 003250-100G
FLUO003250-25GEA 110 EUR
FLUO003250-25G FLUO003250-100G
Hexafluoraceton Sesquihydrat
Hexafluoraceton Sesquihydrat
Formel: C₃F₆O·1.5H₂O
Molecular Weight: 193,05 g/mol
MDL: MFCD00149063
CAS-Nummer: 13098-39-0
EINECS: 211-676-3
UN: 2552
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR