Hexafluoraceton Trihydrat
Lieferant: FLUOROCHEM
Synonyme:
Perfluoroacetone trihydrate
006552-25G
006552-100G
FLUO006552-25GEA
60.9
EUR
FLUO006552-25G
FLUO006552-100G
Hexafluoraceton Trihydrat
Hexafluoraceton Trihydrat
Formel:
C₃F₆O·3H₂O Molecular Weight: 220,07 g/mol |
MDL:
MFCD00149061 CAS-Nummer: 34202-69-2 EINECS: 211-676-3 UN: 2552 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...