Hexafluoraceton Trihydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

139238-25G 139238-5G
SIAL139238-25GEA 73.1 EUR
SIAL139238-25G SIAL139238-5G
Hexafluoraceton Trihydrat, Sigma-Aldrich®
Hexafluoraceton Trihydrat
Formel: C₃F₆O·3H₂O
MDL: MFCD00149061
CAS-Nummer: 34202-69-2
EINECS: 211-676-3
UN: 2552
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR