2-Chlormalondialdehyd 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloromalonaldehyde

330120010 330120100 330120500
ACRO330120010EA 41.8 EUR
ACRO330120010 ACRO330120100 ACRO330120500
2-Chlormalondialdehyd 95%
2-Chlormalondialdehyd
Formel: C₃H₃ClO₂
Molecular Weight: 106.51 g/mol
Schmelzpunkt: 140…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01320489
CAS-Nummer: 36437-19-1

Bestellen

Spezifikation Testergebnisse

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR