2-Chlor-N-(2,6-dimethylphenyl)acetamid

Lieferant: CHEMPUR

Synonyme: N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide

SEW03862.250 SEW03862.1
CHMPSEW03862.250EA 0 EUR
CHMPSEW03862.250 MAYBSEW03862.1
2-Chlor-N-(2,6-dimethylphenyl)acetamid
2-Chlor-N-(2,6-dimethylphenyl)acetamid
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,66 g/mol
MDL: MFCD00000926
CAS-Nummer: 1131-01-7
EINECS: 214-460-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR