2-Chlor-N-(3,5-dimethylphenyl)acetamid

Lieferant: BIONET RESEARCH

Synonyme: N-(Chloroacetyl)-3,5-dimethylaniline

11X-0967.10 11X-0967.1 11X-0967.5 11X-0967.500
BIOT11X-0967.10EA 0 EUR
BIOT11X-0967.10 BIOT11X-0967.1 BIOT11X-0967.5 BIOT11X-0967.500
2-Chlor-N-(3,5-dimethylphenyl)acetamid
2-Chlor-N-(3,5-dimethylphenyl)acetamid
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197.66 g/mol
CAS-Nummer: 174700-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR