Kaliumethylmalonat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Monoethyl malonate potassium salt, Ethyl potassium malonate

248080250 248081000 248085000
ACRO248080250EA 45.7 EUR
ACRO248080250 ACRO248081000 ACRO248085000
Kaliumethylmalonat 98%
Kaliumethylmalonat
Formel: C₅H₇KO₄
Molecular Weight: 170,21 g/mol
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035603
CAS-Nummer: 6148-64-7
EINECS: 228-156-7

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR