Kaliumethylmalonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Monoethyl malonate potassium salt, Ethyl potassium malonate
M-Clarity™ Qualitätsstufe = MQ200
360899-100G
360899-25G
SIAL360899-100GEA
235
EUR
SIAL360899-100G
SIAL360899-25G
Kaliumethylmalonat, Sigma-Aldrich®
Kaliumethylmalonat
Formel:
C₅H₇KO₄ Molecular Weight: 170.21 g/mol |
MDL:
MFCD00035603 CAS-Nummer: 6148-64-7 EINECS: 228-156-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...