Kaliummethylmalonat 98%

Lieferant: Thermo Scientific

Synonyme: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

L20429.14 L20429.22
L20429.14EA 42.6 EUR
L20429.14 L20429.22
Kaliummethylmalonat 98%
Kaliummethylmalonat

Purity: 98% CAS No.: 38330-80-2 Molecular Formula: CH3O2CCH2CO2K Formula Weight: 156.18 Melting Point: ca 190° dec. MDL No.: MFCD00014021

Formel: C₄H₅KO₄
Molecular Weight: 156,18 g/mol
Schmelzpunkt: ∼ 190 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014021
CAS-Nummer: 38330-80-2
EINECS: 253-886-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR