Di-tert-butylazodicarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Di-tert-butyl diazene-1,2-dicarboxylate

M-Clarity™ Qualitätsstufe = MQ200

135992-25G 135992-5G
SIAL135992-25GEA 167 EUR
SIAL135992-25G SIAL135992-5G
Di-tert-butylazodicarboxylat, Sigma-Aldrich®
Di-tert-butylazodicarboxylat
Formel: C₁₀H₁₈N₂O₄
Molecular Weight: 230,26 g/mol
MDL: MFCD00015001
CAS-Nummer: 870-50-8
EINECS: 212-796-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR