Diisopropylazodicarboxylat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Diisopropyl diazene-1,2-dicarboxylate
M-Clarity™ Qualitätsstufe = MQ200
225541-100G
225541-5G
SIAL225541-100GEA
157
EUR
SIAL225541-100G
SIAL225541-5G
Diisopropylazodicarboxylat, Sigma-Aldrich®
Diisopropylazodicarboxylat
Formel:
C₈H₁₄N₂O₄ Molecular Weight: 202,21 g/mol |
MDL:
MFCD00008875 CAS-Nummer: 2446-83-5 EINECS: 219-502-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...