(2-Naphthoxy)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Naphthoxyacetic acid

M-Clarity™ Qualitätsstufe = MQ200

N3019-25G N3019-500G
SIALN3019-25GEA 39.5 EUR
SIALN3019-25G SIALN3019-500G
(2-Naphthoxy)essigsäure, Sigma-Aldrich®
(2-Naphthoxy)essigsäure
Formel: C₁₂H₁₀O₃
Molecular Weight: 202.21 g/mol
MDL: MFCD00004066
CAS-Nummer: 120-23-0
EINECS: 204-380-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR