(2-Naphthoxy)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Naphthoxyacetic acid
M-Clarity™ Qualitätsstufe = MQ100
N3405-100G
N3405-500G
SIALN3405-100GEA
52
EUR
SIALN3405-100G
SIALN3405-500G
(2-Naphthoxy)essigsäure, Sigma-Aldrich®
(2-Naphthoxy)essigsäure
Formel:
C₁₂H₁₀O₃ Molecular Weight: 202,21 g/mol |
MDL:
MFCD00004066 CAS-Nummer: 120-23-0 EINECS: 204-380-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...