N-(3-Pyridyl)thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Pyridyl)-2-thiourea, 1-(Pyridin-3-yl)thiourea

018987-1G 018987-5G
FLUO018987-1GEA 0 EUR
FLUO018987-1G FLUO018987-5G
N-(3-Pyridyl)thioharnstoff
N-(3-Pyridyl)thioharnstoff
Formel: C₆H₇N₃S
Molecular Weight: 153.21 g/mol
MDL: MFCD00014635
CAS-Nummer: 30162-37-9
EINECS: 250-075-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR