N-(4-Pyridyl)thioharnstoff

Lieferant: FLUOROCHEM

CFF189.2.5 CFF189.10 017258-2.5G 017258-10G
CHMPCFF189.2.5EA 268 EUR
CHMPCFF189.2.5 CHMPCFF189.10 FLUO017258-2.5G FLUO017258-10G
N-(4-Pyridyl)thioharnstoff
N-(4-Pyridyl)thioharnstoff
Formel: C₆H₇N₃S
Molecular Weight: 153.21 g/mol
MDL: MFCD00068177
CAS-Nummer: 164670-44-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR