N-(4-Pyridyl)thioharnstoff
Lieferant: Thermo Scientific
SEW02754DA
SEW02754EA
MAYBSEW02754DAEA
0
EUR
MAYBSEW02754DA
MAYBSEW02754EA
N-(4-Pyridyl)thioharnstoff
N-(4-Pyridyl)thioharnstoff
Formel:
C₆H₇N₃S Molecular Weight: 153.21 g/mol |
MDL:
MFCD00068177 CAS-Nummer: 164670-44-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...