N-Methyl-2,2,2-trifluoracetamid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Methyltrifluoroacetamide, 2,2,2-Trifluoro-N-methylacetamide
B23851.06
B23851.14
B23851.06EA
42.9
EUR
B23851.06
B23851.14
N-Methyl-2,2,2-trifluoracetamid ≥98%
N-Methyl-2,2,2-trifluoracetamid
Purity: 98+%
CAS No.: 815-06-5
Molecular Formula: C3H4F3NO
Formula Weight: 127.07
Flash Point: 73°(163°F)
Melting Point: 49-52°
Boiling Point: 156-157°
Beilstein Reference: 1703392
MDL No.: MFCD00009670
Formel:
C₃H₄F₃NO Molecular Weight: 127,07 g/mol Siedepunkt: 156…157 °C Schmelzpunkt: 49…52 °C Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009670 CAS-Nummer: 815-06-5 EINECS: 212-417-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...