Tris(2-aminoethyl)amin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N1,N1-Bis(2-aminoethyl)ethane-1,2-diamine, TREN, 2,2',2''-Triaminotriethylamine

B21789.06 B21789.14 B21789.22 B21789.36
B21789.06EA 30.4 EUR
B21789.06 B21789.14 B21789.22 B21789.36
Tris(2-aminoethyl)amin 97%
Tris(2-aminoethyl)amin

Purity: 97% CAS No.: 4097-89-6 Molecular Formula: C6H18N4 Formula Weight: 146.24 Flash Point: 110°(230°F) Refractive Index: 1.4969 Odour: Amine-like pH: 11.9 Boiling Point: 102°/0.05mm Beilstein Reference: 1739626 UN No.: 2922 MDL No.: MFCD00008177

Formel: C₆H₁₈N₄
Molecular Weight: 146.24 g/mol
Siedepunkt: 102 °C (0.05 mmHg)
Dichte: 0.978
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008177
CAS-Nummer: 4097-89-6
EINECS: 223-857-4
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR