Tris(2-aminoethyl)amin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N1-Bis(2-aminoethyl)ethane-1,2-diamine, TREN, 2,2',2''-Triaminotriethylamine

M-Clarity™ Qualitätsstufe = MQ200

225630-100ML 225630-10ML
SIAL225630-100MLEA 213 EUR
SIAL225630-100ML SIAL225630-10ML
Tris(2-aminoethyl)amin, Sigma-Aldrich®
Tris(2-aminoethyl)amin
Formel: C₆H₁₈N₄
Molecular Weight: 146,24 g/mol
MDL: MFCD00008177
CAS-Nummer: 4097-89-6
EINECS: 223-857-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR