Triethylentetramin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

90460-10ML 90460-50ML
SIAL90460-10MLEA 182 EUR
SIAL90460-10ML SIAL90460-50ML
Triethylentetramin, Sigma-Aldrich®
Triethylentetramin
Formel: C₆H₁₈N₄
MDL: MFCD00008169
CAS-Nummer: 112-24-3
EINECS: 203-950-6
UN: 2259
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR