Dibenzylazodicarboxylat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dibenzyl diazene-1,2-dicarboxylate

L19347.06 L19347.14
L19347.06EA 53.3 EUR
L19347.06 L19347.14
Dibenzylazodicarboxylat 96%
Dibenzylazodicarboxylat
Formel: C₁₆H₁₄N₂O₄
Molecular Weight: 298,3 g/mol
Schmelzpunkt: 43…47 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00016737
CAS-Nummer: 2449-05-0
EINECS: 219-508-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR