Dibenzylazodicarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dibenzyl diazene-1,2-dicarboxylate

M-Clarity™ Qualitätsstufe = MQ100

166804-25G 166804-5G
SIAL166804-25GEA 466 EUR
SIAL166804-25G SIAL166804-5G
Dibenzylazodicarboxylat, Sigma-Aldrich®
Dibenzylazodicarboxylat
Formel: C₁₆H₁₄N₂O₄
Molecular Weight: 298,3 g/mol
MDL: MFCD00016737
CAS-Nummer: 2449-05-0
EINECS: 219-508-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR